Anti-VN1R5 Rabbit Polyclonal Antibody
Catalog # 77076-242
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:G-protein coupled receptor GPCR26
- Antigen Symbol:VN1R5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q7Z5H4
- Antigen Synonyms:V1r like receptor 5|VN1R5|V1r-like receptor 5|V1RL5|hGPCR26|VN1R5_HUMAN|Vomeronasal 1 receptor 5|hGPCR 26|V1R like 5
- Storage Buffer:Supplied in Phosphate Buffered Saline (without Mg2+ and Ca2+), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular Weight:40 kDa
- Storage Temperature:Shipped at 4 ℃. Upon delivery aliquot and store at –20 ℃. Avoid freeze/thaw cycles.
- Shipping Temperature:Shipped on blue ice at 4 ℃
- Immunogen:Synthetic peptide derived from human VN1R5 (amino acids 44 - 93)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Cat. No.:77076-242
- Packaging:Plastic vial
Specifications
About this item
Rabbit polyclonal antibody to VN1R5 for IF and ELISA with samples derived from Human.
- Validated Applications: IF, ELISA
- Recommended Dilutions: IF: 1:100 to 1:500, ELISA: 1:20000
Type: Primary
Antigen: VN1R5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human