Anti-Exodus 2 Rabbit Polyclonal Antibody
Catalog # 77076-646
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:6Ckine
- Antigen Symbol:Exodus 2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# O00585
- Antigen Synonyms:C-C motif chemokine 21|CKb 9|CCL 21|Chemokine (C-C motif) ligand 21|Beta chemokine exodus 2|Beta-chemokine exodus-2|CCL21|Beta chemokine exodus-2|CCL21_HUMAN
- Storage Buffer:Supplied in Phosphate Buffered Saline (without Mg2+ and Ca2+), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular Weight:14 kDa
- Storage Temperature:Shipped at 4 ℃. Upon delivery aliquot and store at –20 ℃. Avoid freeze/thaw cycles.
- Shipping Temperature:Shipped on blue ice at 4 ℃
- Immunogen:Synthetic peptide derived from the Internal region of human CCL21 (amino acids 71 - 120)
- Tested Applications:IHC
- Purification:The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific peptide
- Cat. No.:77076-646
- Packaging:Plastic vial
Specifications
About this item
Rabbit polyclonal antibody to CCL21 for IHC and ELISA with samples derived from Human.
- Validated Applications: IHC, ELISA
- Recommended Dilutions: IHC-P: 1:50 to 1:300, ELISA: 1:10000
Type: Primary
Antigen: Exodus 2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human