Anti-Ataxin 2-like Rabbit Polyclonal Antibody
Catalog # 77077-430
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:Ataxin-2 domain protein
- Antigen Symbol:Ataxin 2-like
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q8WWM7
- Antigen Synonyms:Ataxin 2 like protein|Ataxin 2 related protein|A2RP|Ataxin 2 domain protein|Ataxin 2-like|A2D|Ataxin 2 like|A2LP|A2LG
- Storage Buffer:Supplied in Phosphate Buffered Saline (without Mg2+ and Ca2+), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular Weight:113 kDa
- Storage Temperature:Shipped at 4 ℃. Upon delivery aliquot and store at –20 ℃. Avoid freeze/thaw cycles.
- Shipping Temperature:Shipped on blue ice at 4 ℃
- Immunogen:Synthetic peptide derived from human ATXN2L (amino acids 571 - 620)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Cat. No.:77077-430
- Packaging:Plastic vial
Specifications
About this item
Rabbit polyclonal antibody to ATXN2L for WB and ELISA with samples derived from Human.
- Validated Applications: WB, ELISA
- Recommended Dilutions: WB: 1:500 to 1:1000, ELISA: 1:1000
Type: Primary
Antigen: Ataxin 2-like
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human