Anti-CLCNKA Rabbit Polyclonal Antibody
77069-170
- Antibody Type:Primary
- Antigen Name:C75963
- Antigen Symbol:CLCNKA
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P51800
- Antigen Synonyms:hClC Ka|ClC K1|CLCK1|CLCNKA|ClC-K1|Chloride channel kidney A|Chloride channel protein ClC Ka|Chloride channel protein ClC-Ka|Chloride channel Ka
- Storage Buffer:Supplied in Phosphate Buffered Saline (without Mg2+ and Ca2+), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular Weight:75 kDa
- Storage Temperature:Shipped at 4 ℃. Upon delivery aliquot and store at –20 ℃. Avoid freeze/thaw cycles.
- Shipping Temperature:Shipped on blue ice at 4 ℃
- Immunogen:Synthetic peptide derived from human CLCNKA (amino acids 567 - 616)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Cat. No.:77069-170
- Packaging:Plastic vial
Rabbit polyclonal antibody to CLCNKA for WB and ELISA with samples derived from Human.
- Validated Applications: WB, ELISA
- Recommended Dilutions: WB: 1:500 to 1:1000, ELISA: 1:10000
Type: Primary
Antigen: CLCNKA
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human