Anti-MRPL18 Rabbit Polyclonal Antibody
77069-266
- Antibody Type:Primary
- Antigen Name:39S ribosomal protein L18
- Antigen Symbol:MRPL18
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q9H0U6
- Antigen Synonyms:39S ribosomal protein L18, mitochondrial precursor|39S ribosomal protein L18 mitochondrial|MRPL18|RM18_HUMAN|MRP-L18|Mitochondrial ribosomal protein L18|HSPC071|L18mt|Mitochondrial ribosomal protein L18 (MRPL18 protein)
- Storage Buffer:Supplied in Phosphate Buffered Saline (without Mg2+ and Ca2+), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular Weight:20 kDa
- Storage Temperature:Shipped at 4 ℃. Upon delivery aliquot and store at –20 ℃. Avoid freeze/thaw cycles.
- Shipping Temperature:Shipped on blue ice at 4 ℃
- Immunogen:Synthetic peptide derived from human MRPL18 (amino acids 1 - 50)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Cat. No.:77069-266
- Packaging:Plastic vial
Rabbit polyclonal antibody to MRPL18 for WB and ELISA with samples derived from Human.
- Validated Applications: WB, ELISA
- Recommended Dilutions: WB: 1:500 to 1:1000, ELISA: 1:1000
Type: Primary
Antigen: MRPL18
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human