Anti-OR2AK2 Rabbit Polyclonal Antibody
Catalog # 77078-980
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:O2AK2_HUMAN
- Antigen Symbol:OR2AK2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q8NG84
- Antigen Synonyms:Olfactory receptor OR1-47|Olfactory receptor family 2 subfamily AK member 1 pseudogene|OR2AK1P|Olfactory receptor 2AK1|OR1 47|Olfactory receptor 2AK2|OR2AK2|Olfactory receptor family 2 subfamily AK member 2
- Storage Buffer:Supplied in Phosphate Buffered Saline (without Mg2+ and Ca2+), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular Weight:36 kDa
- Storage Temperature:Shipped at 4 ℃. Upon delivery aliquot and store at –20 ℃. Avoid freeze/thaw cycles.
- Shipping Temperature:Shipped on blue ice at 4 ℃
- Immunogen:Synthetic peptide derived from human OR2AK2 (amino acids 241 - 290)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Cat. No.:77078-980
- Packaging:Plastic vial
Specifications
About this item
Rabbit polyclonal antibody to OR2AK2 for IF and ELISA with samples derived from Human.
- Validated Applications: IF, ELISA
- Recommended Dilutions: IF: 1:100 to 1:500, ELISA: 1:5000
Type: Primary
Antigen: OR2AK2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human