Anti-Glucose Transporter 5 GLUT5 Rabbit Polyclonal Antibody
Catalog # 77077-522
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:Facilitated glucose transporter member 5
- Antigen Symbol:Glucose Transporter 5 GLUT5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P22732
- Antigen Synonyms:Glucose transporte, kidney|GLUT 5|Glucose transporter type 5|GTR5_HUMAN|GLUT-5|Fructose transporter|glucose transporter like protein 5|Glucose Transporter 5 GLUT5|Glucose transporter type 5 small intestine
- Storage Buffer:Supplied in Phosphate Buffered Saline (without Mg2+ and Ca2+), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular Weight:54 kDa
- Storage Temperature:Shipped at 4 ℃. Upon delivery aliquot and store at –20 ℃. Avoid freeze/thaw cycles.
- Shipping Temperature:Shipped on blue ice at 4 ℃
- Immunogen:Synthetic peptide derived from the N - terminal region of human SLC2A5 (amino acids 31 - 80)
- Purification:The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using peptide
- Cat. No.:77077-522
- Packaging:Plastic vial
Specifications
About this item
Rabbit polyclonal antibody to SLC2A5 for WB and ELISA with samples derived from Human.
- Validated Applications: WB, ELISA
- Recommended Dilutions: WB: 1:500 to 1:1000, ELISA: 1:10000
Type: Primary
Antigen: Glucose Transporter 5 GLUT5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human