Anti-ELOVL2 Rabbit Polyclonal Antibody
Catalog # 77065-880
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:3 keto acyl CoA synthase ELOVL2
- Antigen Symbol:ELOVL2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q9NXB9
- Antigen Synonyms:FLJ20334|Elongation of very long chain fatty acids (FEN1/Elo2, SUR4/Elo3, yeast) like 2|Elongation of very long chain fatty acids protein 2|OTTHUMP00000016029|ELOV2_HUMAN|3-keto acyl-CoA synthase Elovl2|SSC2|ELOVL fatty acid elongase 2
- Storage Buffer:Supplied in Phosphate Buffered Saline (without Mg2+ and Ca2+), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular Weight:34 kDa
- Storage Temperature:Shipped at 4 ℃. Upon delivery aliquot and store at –20 ℃. Avoid freeze/thaw cycles.
- Shipping Temperature:Shipped on blue ice at 4 ℃
- Immunogen:Synthetic peptide derived from human ELOVL2 (amino acids 250 - 296)
- Tested Applications:IHC
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Cat. No.:77065-880
- Packaging:Plastic vial
Specifications
About this item
Rabbit polyclonal antibody to ELOVL2 for IHC and ELISA with samples derived from Human.
- Validated Applications: IHC, ELISA
- Recommended Dilutions: IHC: 1:50 to 1:100, ELISA: 1:10000
Type: Primary
Antigen: ELOVL2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human