Anti-ABHD12B Rabbit Polyclonal Antibody
77074-838
- Antibody Type:Primary
- Antigen Name:AB12B_HUMAN
- Antigen Symbol:ABHD12B
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q7Z5M8
- Antigen Synonyms:Abhydrolase domain containing 12B|Bem46l3|Abhydrolase domain-containing protein 12B|c14_5314|ABHD12B|Gm804|C14orf29
- Storage Buffer:Supplied in Phosphate Buffered Saline (without Mg2+ and Ca2+), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular Weight:40 kDa
- Storage Temperature:Shipped at 4 ℃. Upon delivery aliquot and store at –20 ℃. Avoid freeze/thaw cycles.
- Shipping Temperature:Shipped on blue ice at 4 ℃
- Immunogen:Synthetic peptide derived from human ABHD12B (amino acids 281 - 330)
- Tested Applications:IHC
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Cat. No.:77074-838
- Packaging:Plastic vial
Rabbit polyclonal antibody to ABHD12B for IHC and ELISA with samples derived from Human.
- Validated Applications: IHC, ELISA
- Recommended Dilutions: IHC: 1:50 to 1:100, ELISA: 1:20000
Type: Primary
Antigen: ABHD12B
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human