Anti-Acyl-coenzyme A Thioesterase 4 Rabbit Polyclonal Antibody
Catalog # 77068-252
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:ACOT4
- Antigen Symbol:Acyl-coenzyme A Thioesterase 4
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q8N9L9
- Antigen Synonyms:Acyl-coenzyme A Thioesterase 4|Acyl-CoA thioesterase 4|Peroxisomal acyl coenzyme A thioester hydrolase Ib|ACOT4_HUMAN|Peroxisomal long-chain acyl-CoA thioesterase Ib|PTE-2b|Peroxisomal acyl CoA thioesterase 2B|Pte1b|PTE-Ib
- Storage Buffer:Supplied in Phosphate Buffered Saline (without Mg2+ and Ca2+), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular Weight:46 kDa
- Storage Temperature:Shipped at 4 ℃. Upon delivery aliquot and store at –20 ℃. Avoid freeze/thaw cycles.
- Shipping Temperature:Shipped on blue ice at 4 ℃
- Immunogen:Synthetic peptide derived from human ACOT4 (amino acids 361 - 410)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Cat. No.:77068-252
- Packaging:Plastic vial
Specifications
About this item
Rabbit polyclonal antibody to ACOT4 for WB, IF and ELISA with samples derived from Human.
- Validated Applications: WB, IF, ELISA
- Recommended Dilutions: WB: 1:500 to 1:1000, IF: 1:100 to 1:500, ELISA: 1:20000
Type: Primary
Antigen: Acyl-coenzyme A Thioesterase 4
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human