Anti-PTBP1 Rabbit Monoclonal Antibody [Clone: ARC0783]
Supplier: ANTIBODIES.COM LLC
Certificates
About this item
Rabbit monoclonal [ARC0783] antibody to PTBP1 for WB, IHC, ICC/IF and IP with samples derived from Human, Mouse and Rat.
- Validated Applications: WB, IHC, ICC/IF, IP
- Recommended Dilutions: WB: 1:500 to 1:2,000, IHC: 1:50 to 1:200, ICC/IF: 1:50 to 1:200, IP: 1:500 to 1:1,000
Type: Primary
Antigen: PTBP1
Clonality: Monoclonal
Clone: ARC0783
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Frequently Bought Together