Anti-c-Myc (phospho Thr58) Rabbit Antibody [CLone: ARC1534]
Supplier: ANTIBODIES.COM LLC
Certificates
About this item
Rabbit monoclonal [ARC1534] antibody to c-Myc (phospho Thr58) for WB with samples derived from Human.
- Validated Applications: WB
- Recommended Dilutions: WB: 1:500 to 1:2,000
Type: Primary
Antigen: c-Myc
Clonality: Monoclonal
Clone: ARC1534
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Frequently Bought Together