Anti-Reptin / TIP49B / RUVB2 Rabbit Monoclonal Antibody [Clone: ARC0933]
Supplier: ANTIBODIES.COM LLC
Certificates
About this item
Rabbit monoclonal [ARC0933] antibody to Reptin / TIP49B / RUVB2 for WB with samples derived from Human, Mouse and Rat.
- Validated Applications: WB
- Recommended Dilutions: WB: 1:500 to 1:2,000
Type: Primary
Antigen: Reptin / TIP49B / RUVB2
Clonality: Monoclonal
Clone: ARC0933
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Related Information
Frequently Bought Together