Anti-beta Catenin (phospho Ser33 + Ser37) Rabbit Antibody [CLone: ARC1523]
Supplier: ANTIBODIES.COM LLC
Certificates
About this item
Rabbit monoclonal [ARC1523] antibody to beta Catenin (phospho Ser33 + Ser37) for WB with samples derived from Human.
- Validated Applications: WB
- Recommended Dilutions: WB: 1:500 to 1:2,000
Type: Primary
Antigen: beta Catenin
Clonality: Monoclonal
Clone: ARC1523
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Related Information
Frequently Bought Together