Anti-Otx2 Rabbit Monoclonal Antibody [Clone: ARC0973]
Rabbit monoclonal [ARC0973] antibody to Otx2 for WB with samples derived from Human, Mouse and Rat.
- Validated Applications: WB
- Recommended Dilutions: WB: 1:500 to 1:2,000
Type: Primary
Antigen: Otx2
Clonality: Monoclonal
Clone: ARC0973
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat
Frequently Bought Together