Order Entry
United States
ContactUsLinkComponent
Anti-TAK1 Rabbit Polyclonal Antibody
Anti-TAK1 Rabbit Polyclonal Antibody
Catalog # 77172-856
Supplier:  ANTIBODIES.COM LLC
undefined
Anti-TAK1 Rabbit Polyclonal Antibody
Catalog # 77172-856
Supplier:  ANTIBODIES.COM LLC
Supplier Number:  A305052-100

Specifications

  • Cat. No.:
    77172-856

Specifications

About this item

Rabbit polyclonal antibody to TAK1 for WB and IHC with samples derived from Human, Mouse and Rat.

  • Validated Applications: WB, IHC
  • Recommended Dilutions: WB: 1:1,000, IHC: 1:50

Type: Primary
Antigen: TAK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat