Anti-AHA1 Rat Monoclonal Antibody [Clone: 25F2.D10]
77172-748
- Antibody Type:Primary
- Antigen Name:Activator of 90 kDa heat shock protein ATPase homolog 1
- Antigen Symbol:AHA1
- Clonality:Monoclonal
- Clone:25F2.D10
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rat
- ImmunoChemistry:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG2a kappa
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q8BK64
- Antigen Synonyms:HSPC322|AHA1|C14orf3|Activator of heat shock 90kDa protein ATPase homolog 1|p38|AHSA 1|AHSA1_HUMAN|Ahsa1|AHA 1
- Storage Buffer:Supplied in 20mM Potassium Phosphate, pH 7.2, with 0.15M Sodium Chloride
- Molecular Weight:~38 kDa (can run up to 45 kDa on SDS-PAGE)
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze / thaw cycles.
- Concentration:1.59 mg/ml
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Mouse Aha1.
- Tested Applications:IHC, ICC/IF
- Purification:Protein G purification
- Cat. No.:77172-748
- Packaging:Plastic vial
Rat monoclonal [25F2.D10] antibody to AHA1 for WB, IHC, ICC/IF, IP and ELISA with samples derived from Human, Mouse and Rat.
- Validated Applications: WB, IHC, ICC/IF, IP, ELISA
- Recommended Dilutions: WB: 1:1,000, IHC: 1:100, ICC/IF: 1:1,000, IP: 1:1,000
Type: Primary
Antigen: AHA1
Clonality: Monoclonal
Clone: 25F2.D10
Conjugation: Unconjugated
Epitope:
Host: Rat
Isotype: IgG2a kappa
Reactivity: Human, Mouse, Rat