Order Entry
United States
ContactUsLinkComponent
Anti-RICTOR Rabbit Polyclonal Antibody
Anti-RICTOR Rabbit Polyclonal Antibody
Catalog # 77172-632
Supplier:  ANTIBODIES.COM LLC
undefined
Anti-RICTOR Rabbit Polyclonal Antibody
Catalog # 77172-632
Supplier:  ANTIBODIES.COM LLC
Supplier Number:  A304940-100

Specifications

  • Cat. No.:
    77172-632

Specifications

About this item

Rabbit polyclonal antibody to RICTOR for WB and IHC with samples derived from Human and Mouse.

  • Validated Applications: WB, IHC
  • Recommended Dilutions: WB: 1:1,000

Type: Primary
Antigen: RICTOR
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse