Anti-HSP70 Mouse Monoclonal Antibody [Clone: 1H11]
Catalog # 77172-176
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:DnaK type molecular chaperone HSP70 1
- Antigen Symbol:HSP70
- Clonality:Monoclonal
- Clone:1H11
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG1
- Reactivity:Pig,Human,Bovine,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P0DMV8, P0DMV9
- Antigen Synonyms:Heat shock 70 kDa protein 1|FLJ54303|FLJ75127|FLJ54408|Epididymis secretory protein Li 103|Heat shock 70 kDa protein 1A/1B|Heat shock 70 kDa protein 1/2|FLJ54370|FLJ54392
- Storage Buffer:Supplied in Phosphate Buffered Saline, pH 7.4, with 50% Glycerol and 0.09% Sodium Azide
- Molecular Weight:~70 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze / thaw cycles.
- Concentration:0.5 mg/ml
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Human native HSP70 protein.
- Tested Applications:ICC/IF, FACS
- Purification:Protein G purification
- Cat. No.:77172-176
- Packaging:Plastic vial
Specifications
About this item
Mouse monoclonal [1H11] antibody to HSP70 for WB, ICC/IF, Flow Cytometry and FACS with samples derived from Human, Mouse, Rat, Bovine and Porcine.
- Validated Applications: WB, ICC/IF, Flow Cytometry, FACS
- Recommended Dilutions: WB: 1:1,000, ICC/IF: 1:100, FACS: 1:250
Type: Primary
Antigen: HSP70
Clonality: Monoclonal
Clone: 1H11
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human, Mouse, Rat, Bovine, Porcine