Anti-5-Carboxylcytosine (5-caC) Rabbit Recombinant Antibody [Clone: RM462]
Catalog # 76833-850
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:5-carboxylcytosine (5-caC)
- Antigen Symbol:5-Carboxylcytosine (5-caC)
- Clonality:Recombinant
- Clone:RM462
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Vertebrates
- Size:50 µg
- Form:Liquid
- Antigen Synonyms:5-carboxylcytosine (5-caC)
- Storage Buffer:Supplied in Phosphate Buffered Saline with 50% Glycerol, 1% BSA, and 0.09% Sodium Azide
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze / thaw cycles.
- Concentration:1 mg/ml
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:KLH-conjugated 5-Carboxylcytosine.
- Tested Applications:Dot Blot
- Purification:Protein A chromatography
- Cat. No.:76833-850
- Packaging:Plastic vial
Specifications
About this item
Rabbit monoclonal [RM462] antibody to 5-Carboxylcytosine (5-caC) for Dot Blot and ELISA with samples derived from All Vertebrates.
- Validated Applications: Dot Blot, ELISA
- Recommended Dilutions: Dot Blot: 0.1 to 1 µg/ml, ELISA: 0.2 to 2 µg/ml
Type: Primary
Antigen: 5-Carboxylcytosine (5-caC)
Clonality: Recombinant
Clone: RM462
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Vertebrates