Anti-Mouse IgG2c Goat Polyclonal Antibody (Biotin)
76833-206
- Antibody Type:Secondary
- Antigen Symbol:Mouse IgG2c
- Clonality:Polyclonal
- Conjugation:Biotin
- ELISA:Yes
- Host:Goat
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Mouse
- Western Blot:Yes
- Size:1 mg
- Form:Liquid
- Antibody Fragment:Whole Molecule
- Storage Buffer:Supplied in Phosphate Buffered Saline with 0.2% BSA and 0.09% Sodium Azide
- Storage Temperature:Shipped at 4 °C. Store at 4 °C. Do not freeze!
- Concentration:1 mg/ml
- Shipping Temperature:Shipped on blue ice at 4 °C
- Tested Applications:IHC, ICC
- Purification:Antigen affinity purification
- Cat. No.:76833-206
- Packaging:Plastic vial
Goat anti-mouse IgG2c (Biotin) secondary antibody for WB, IHC, ICC and ELISA with samples derived from Mouse.
- Validated Applications: WB, IHC, ICC, ELISA
- Recommended Dilutions: ELISA: 1:10,000 to 1:200,000, ICC: 1:100 to 1:500, IHC: 1:250 to 1:2,500, WB: 1:10,000 to 1:200,000
Type: Secondary
Antigen: Mouse IgG2c
Clonality: Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Goat
Isotype: IgG
Reactivity: Mouse