Anti-Human IgG H&L Rabbit Polyclonal Antibody (AP (Alkaline Phosphatase) Then)
Catalog # 76835-346
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Secondary
- Antigen Symbol:Human IgG H&L
- Clonality:Polyclonal
- Conjugation:AP (Alkaline Phosphatase)
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:1 mg
- Form:Liquid
- Antibody Fragment:Whole Molecule
- Storage Buffer:Supplied in 50 mM HEPES, pH 7.1, with 0.1 M Sodium Chloride, 1 mM Magnesium Chloride, 0.1 mM Zinc Chloride, 0.2% BSA, and 0.09% Sodium Azide
- Storage Temperature:Store in the dark at 4 °C. Avoid prolonged exposure to light. Do not freeze!
- Concentration:1 mg/ml
- Shipping Temperature:Shipped on blue ice at 4 °C
- Tested Applications:IHC, ICC
- Purification:Antigen affinity purification
- Cat. No.:76835-346
- Packaging:Plastic vial
Specifications
About this item
Rabbit anti-human IgG H&L (AP) secondary antibody for WB, IHC, ICC and ELISA with samples derived from Human.
- Validated Applications: WB, IHC, ICC, ELISA
- Recommended Dilutions: WB: 1:5,000 to 1:50,000, IHC: 1:100 to 1:1,000, ICC: 1:100 to 1:1,000, ELISA: 1:5,000 to 1:50,000
Type: Secondary
Antigen: Human IgG H&L
Clonality: Polyclonal
Clone:
Conjugation: AP (Alkaline Phosphatase)
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human