Order Entry
United States
ContactUsLinkComponent
Anti-Goat IgM Heavy Chain Rabbit Polyclonal Antibody
Anti-Goat IgM Heavy Chain Rabbit Polyclonal Antibody
Catalog # 76833-324
Supplier:  ANTIBODIES.COM LLC
Anti-Goat IgM Heavy Chain Rabbit Polyclonal Antibody
Catalog # 76833-324
Supplier:  ANTIBODIES.COM LLC
Supplier Number:  A300693-1

Specifications

  • Cat. No.:
    76833-324

Specifications

About this item

Rabbit anti-goat IgM heavy chain secondary antibody for WB, IHC, ICC and ELISA with samples derived from Goat.

  • Validated Applications: WB, IHC, ICC, ELISA
  • Recommended Dilutions: ELISA: 1:1,000 to 1:30,000; ELISA (capture antibody): 1:100 to 1:500, ICC: 1:200 to 1:2,000, IHC: 1:200 to 1:2,000, WB: 1:1,000 to 1:30,000

Type: Secondary
Antigen: Goat IgM Heavy Chain
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Goat