Order Entry
United States
ContactUsLinkComponent
Anti-Granzyme B Rabbit Recombinant Antibody [Clone: RM441]
Anti-Granzyme B Rabbit Recombinant Antibody [Clone: RM441]
Catalog # 76833-756
Supplier:  Antibodies.com
Anti-Granzyme B Rabbit Recombinant Antibody [Clone: RM441]
Catalog # 76833-756
Supplier:  Antibodies.com
Supplier Number:  A290858-100

Specifications

  • Cat. No.:
    76833-756

Specifications

About this item

Recombinant rabbit monoclonal [RM441] antibody to Granzyme B for IHC with samples derived from Human.

  • Validated Applications: IHC
  • Recommended Dilutions: IHC: 1:100 to 1:400

Type: Primary
Antigen: Granzyme B
Clonality: Recombinant
Clone: RM441
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human