Anti-ZNF562 Mouse Monoclonal Antibody [Clone: PCRP-ZNF562-1A1]
Catalog # 76954-666
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Symbol:ZNF562
- Clonality:Monoclonal
- Clone:PCRP-ZNF562-1A1
- Conjugation:Unconjugated
- ELISA:Yes
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG1
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q6V9R5
- Storage Buffer:Supplied in 10mM Phosphate Buffered Saline; without Sodium Azide and carrier free
- Molecular Weight:48.5 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze / thaw cycles.
- Concentration:1 mg/ml
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant full-length human ZNF562 protein.
- Purification:Protein A/G chromatography
- Cat. No.:76954-666
- Packaging:Plastic vial
Specifications
About this item
Mouse monoclonal [PCRP-ZNF562-1A1] antibody to ZNF562 for ELISA, IP, Flow Cytometry and IF with samples derived from Human.
- Validated Applications: ELISA, IP, Flow Cytometry, IF
- Recommended Dilutions: IP: 1 to 2µg / 100 to 500µg proteins, Flow Cytometry: 0.5 to 2 µg/million cells, IF: 0.5 to 2 µg/ml
Type: Primary
Antigen: ZNF562
Clonality: Monoclonal
Clone: PCRP-ZNF562-1A1
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human