Order Entry
United States
ContactUsLinkComponent
Anti-ZNF81 Mouse Monoclonal Antibody [Clone: PCRP-ZNF81-2G2]
Anti-ZNF81 Mouse Monoclonal Antibody [Clone: PCRP-ZNF81-2G2]
Catalog # 76965-378
Supplier:  ANTIBODIES.COM LLC
undefined
Anti-ZNF81 Mouse Monoclonal Antibody [Clone: PCRP-ZNF81-2G2]
Catalog # 76965-378
Supplier:  ANTIBODIES.COM LLC
Supplier Number:  A278245-100

Specifications

  • Cat. No.:
    76965-378

Specifications

About this item

Mouse monoclonal [PCRP-ZNF81-2G2] antibody to ZNF81 for ELISA, WB, IP, Flow Cytometry and IF with samples derived from Human.

  • Validated Applications: ELISA, WB, IP, Flow Cytometry, IF
  • Recommended Dilutions: WB: 1 to 2 µg/ml, IP: 1 to 2µg / 100 to 500µg proteins, Flow Cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ml

Type: Primary
Antigen: ZNF81
Clonality: Monoclonal
Clone: PCRP-ZNF81-2G2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a
Reactivity: Human