Order Entry
United States
ContactUsLinkComponent
Anti-Ornithine Decarboxylase Mouse Recombinant Antibody [Clone: rODC1/487]
Anti-Ornithine Decarboxylase Mouse Recombinant Antibody [Clone: rODC1/487]
Catalog # 76955-146
Supplier:  ANTIBODIES.COM LLC
Anti-Ornithine Decarboxylase Mouse Recombinant Antibody [Clone: rODC1/487]
Catalog # 76955-146
Supplier:  ANTIBODIES.COM LLC
Supplier Number:  A277906-100

Specifications

  • Cat. No.:
    76955-146

Specifications

About this item

Recombinant mouse monoclonal [rODC1/487] antibody to Ornithine Decarboxylase for ELISA and WB with samples derived from Human, Mouse and Rat.

  • Validated Applications: ELISA, WB
  • Recommended Dilutions: WB: 1 to 2 µg/ml

Type: Primary
Antigen: Ornithine Decarboxylase
Clonality: Recombinant
Clone: rODC1/487
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human, Mouse, Rat