Order Entry
United States
ContactUsLinkComponent
Anti-Myeloperoxidase Mouse Monoclonal Antibody [Clone: MPO/7116]
Anti-Myeloperoxidase Mouse Monoclonal Antibody [Clone: MPO/7116]
Catalog # 76960-202
Supplier:  ANTIBODIES.COM LLC
undefined
Anti-Myeloperoxidase Mouse Monoclonal Antibody [Clone: MPO/7116]
Catalog # 76960-202
Supplier:  ANTIBODIES.COM LLC
Supplier Number:  A277714-100

Specifications

  • Cat. No.:
    76960-202

Specifications

About this item

Mouse monoclonal [MPO/7116] antibody to Myeloperoxidase for IHC-P with samples derived from Human.

  • Validated Applications: IHC-P
  • Recommended Dilutions: IHC-P: 1 to 2 µg/ml

Type: Primary
Antigen: Myeloperoxidase
Clonality: Monoclonal
Clone: MPO/7116
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human