Anti-GTF2A1 Mouse Monoclonal Antibody [Clone: PCRP-GTF2A1-1F2]
Catalog # 76963-428
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:General transcription factor IIA 1 19/37kDa
- Antigen Symbol:GTF2A1
- Clonality:Monoclonal
- Clone:PCRP-GTF2A1-1F2
- Conjugation:Unconjugated
- ELISA:Yes
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- ImmunoPrecipitation:Yes
- Isotype:IgG2b
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P52655
- Antigen Synonyms:TF2AA_HUMAN|Glucose regulated protein 58kD pseudogene|General transcription factor IIA subunit 1|TFIIA 42|TFIIA alpha p55, isoform 1|General transcription factor IIA1|TFIIA alpha p55|TFIIA p19 subunit|GTF2A1
- Storage Buffer:Supplied in 10mM Phosphate Buffered Saline with 0.05% BSA and 0.05% Sodium Azide
- Molecular Weight:41.51 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze / thaw cycles.
- Concentration:200 µg/ml
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant full-length human GTF2A1 protein.
- Purification:Protein A/G chromatography
- Cat. No.:76963-428
- Packaging:Plastic vial
Specifications
About this item
Mouse monoclonal [PCRP-GTF2A1-1F2] antibody to GTF2A1 for ELISA, IP, Flow Cytometry and IF with samples derived from Human.
- Validated Applications: ELISA, IP, Flow Cytometry, IF
- Recommended Dilutions: IP: 1 to 2µg / 100 to 500µg proteins, Flow Cytometry: 1 to 2 µg/million cells, IF: 1 to 2 µg/ml
Type: Primary
Antigen: GTF2A1
Clonality: Monoclonal
Clone: PCRP-GTF2A1-1F2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2b
Reactivity: Human