Anti-TOM1L1 Mouse Monoclonal Antibody [Clone: TOM1L1/4690]
Catalog # 76951-222
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:2310045L10Rik
- Antigen Symbol:TOM1L1
- Clonality:Monoclonal
- Clone:TOM1L1/4690
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# O75674
- Antigen Synonyms:Target of myb1 (chicken) homolog like 1|Target of Myb like protein 1|OK/KNS CL.3|Src-activating and signaling molecule protein|C80573|Src activating and signaling molecule protein|SRCASM|Target of Myb-like protein 1|Src activating and signaling molecule
- Storage Buffer:Supplied in 10mM Phosphate Buffered Saline with 0.05% BSA and 0.05% Sodium Azide
- Molecular Weight:53 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze / thaw cycles.
- Concentration:200 µg/ml
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Recombinant full-length human TOM1L1 protein.
- Tested Applications:IHC-P
- Purification:Protein A/G chromatography
- Cat. No.:76951-222
- Packaging:Plastic vial
Specifications
About this item
Mouse monoclonal [TOM1L1/4690] antibody to TOM1L1 for IHC-P with samples derived from Human.
- Validated Applications: IHC-P
- Recommended Dilutions: IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: TOM1L1
Clonality: Monoclonal
Clone: TOM1L1/4690
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG
Reactivity: Human