Anti-EpCAM Mouse Monoclonal Antibody [Clone: GZ-20]
Supplier: ANTIBODIES.COM LLC
Certificates
About this item
Mouse monoclonal [GZ-20] antibody to EpCAM for Flow Cytometry, IF, WB and IP with samples derived from Rat.
- Validated Applications: Flow Cytometry, IF, WB, IP
- Recommended Dilutions: Flow Cytometry: 5 to 10 µl / million cells, IF: 1:50 to 1:100, WB: 1:100 to 1:200, IP: 5 to 10 µl / 500µg protein lysate
Type: Primary
Antigen: EpCAM
Clonality: Monoclonal
Clone: GZ-20
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Rat
Specifications
- Catalog No:
- 76949-458
- 76952-438
- Antigen Symbol:
- EpCAM
- EpCAM
- Antigen Name:
- 17 1A
- 17 1A
- Antigen Synonyms:
- CD326|Antigen identified by monoclonal|Cell surface glycoprotein Trop 1|AUA1|Cell surface glycoprotein Trop 2|CD326 antigen|Adenocarcinoma-associated antigen|Adenocarcinoma associated antigen|323/A3
- CD326|Antigen identified by monoclonal|Cell surface glycoprotein Trop 1|AUA1|Cell surface glycoprotein Trop 2|CD326 antigen|Adenocarcinoma-associated antigen|Adenocarcinoma associated antigen|323/A3
- Antibody Type:
- Primary
- Primary
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Unconjugated
- Unconjugated
- Clone:
- GZ-20
- GZ-20
- Reactivity:
- Rat
- Rat
- Host:
- Mouse
- Mouse
- Gene ID:
- UniprotID# O55159
- UniprotID# O55159
- Isotype:
- IgG1 kappa
- IgG1 kappa
- Western Blot:
- Yes
- Yes
- ImmunoPrecipitation:
- Yes
- Yes
- ImmunoFluorescence:
- Yes
- Yes
- Flow Cytometry:
- Yes
- Yes
- Immunogen:
- Rat small intestine cells.
- Rat small intestine cells.
- Form:
- Liquid
- Liquid
- Molecular Weight:
- 40-43 kDa
- 40-43 kDa
- Storage Buffer:
- Bioreactor concentrate with 0.05% Sodium Azide
- Bioreactor concentrate with 0.05% Sodium Azide
- Storage Temperature:
- Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze / thaw cycles.
- Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze / thaw cycles.
- Shipping Temperature:
- Shipped on blue ice at 4 °C
- Shipped on blue ice at 4 °C
Specifications
Frequently Bought Together