Anti-Melanoma Associated Antigen KBA.62 Mouse Monoclonal Antibody [Clone: KBA.62]
Catalog # 76965-544
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:Melanoma Associated Antigen KBA.62
- Antigen Symbol:Melanoma Associated Antigen KBA.62
- Clonality:Monoclonal
- Clone:KBA.62
- Conjugation:Unconjugated
- Host:Mouse
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG1 kappa
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Antigen Synonyms:Melanoma Associated Antigen KBA.62
- Storage Buffer:Supplied in 10mM Phosphate Buffered Saline; without Sodium Azide and carrier free
- Molecular Weight:140 kDa / 135 kDa / 128 kDa / two weak bands of 88 kDa / 73 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze / thaw cycles.
- Concentration:1 mg/ml
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Human KAL cells derived from lymph node metastasis of malignant melanoma.
- Tested Applications:IHC-P
- Purification:Protein A/G chromatography
- Cat. No.:76965-544
- Packaging:Plastic vial
Specifications
About this item
Mouse monoclonal [KBA.62] antibody to Melanoma Associated Antigen KBA.62 for IF and IHC-P with samples derived from Human.
- Validated Applications: IF, IHC-P
- Recommended Dilutions: IF: 1 to 2 µg/ml, IHC-P: 1 to 2 µg/ml
Type: Primary
Antigen: Melanoma Associated Antigen KBA.62
Clonality: Monoclonal
Clone: KBA.62
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human