Anti-MHC Class II Mouse Monoclonal Antibody [Clone: MK-D6]
Catalog # 76964-428
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:D6S221E
- Antigen Symbol:MHC Class II
- Clonality:Monoclonal
- Clone:MK-D6
- Conjugation:Unconjugated
- Flow Cytometry:Yes
- Host:Mouse
- ImmunoFluorescence:Yes
- Isotype:IgG2a kappa
- Reactivity:Mouse
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P01904, P04224
- Antigen Synonyms:DPB1|DMA|DPB1_HUMAN|DRB|DPB 1|DP beta 1 chain|DP(W4) beta chain|DP beta 1|DMB
- Storage Buffer:Supplied in 10mM Phosphate Buffered Saline; without Sodium Azide and carrier free
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze / thaw cycles.
- Concentration:1 mg/ml
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Spleen cells from B10.D2 mice.
- Tested Applications:Functional Studies
- Purification:Protein A/G chromatography
- Cat. No.:76964-428
- Packaging:Plastic vial
Specifications
About this item
Mouse monoclonal [MK-D6] antibody to MHC Class II for Functional Studies, Flow Cytometry and IF with samples derived from Mouse.
- Validated Applications: Functional Studies, Flow Cytometry, IF
- Recommended Dilutions: Flow Cytometry: 0.5 to 1 µg/million cells, IF: 1 to 2 µg/ml
Type: Primary
Antigen: MHC Class II
Clonality: Monoclonal
Clone: MK-D6
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG2a kappa
Reactivity: Mouse