Anti-ARMC6 Rabbit Polyclonal Antibody
Catalog # 77052-908
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:2410153K17Rik
- Antigen Symbol:ARMC6
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q6NXE6
- Antigen Synonyms:MGC19595|MGC67564|R30923_1|OTTMUSP00000025055|RGD1306691|ARMC6|AW554412|Armadillo repeat containing 6|Armadillo repeat containing protein 6
- Storage Buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular Weight:54 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human ARMC6 (amino acids 431-480)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Cat. No.:77052-908
- Packaging:Plastic vial
Specifications
About this item
Rabbit polyclonal antibody to ARMC6 for WB, IF and ELISA with samples derived from Human, Mouse and Rat.
- Validated Applications: WB, IF, ELISA
- Recommended Dilutions: WB: 1:500-1:1000, IF: 1:100-1:500, ELISA: 1:10000
Type: Primary
Antigen: ARMC6
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat