Anti-KDELR2 Rabbit Polyclonal Antibody
77052-812
- Antibody Type:Primary
- Antigen Name:(Lys Asp Glu Leu) endoplasmic reticulum protein retention receptor 2
- Antigen Symbol:KDELR2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P33947
- Antigen Synonyms:ERD 2 like protein|ERD22_HUMAN|ER lumen protein retaining receptor 2|ELP1|ELP-1|ERD2-like protein 1|ERD2.2|ELP 1|ERD2 like protein 1
- Storage Buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular Weight:24 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human ERD22 (amino acids 81-130)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Cat. No.:77052-812
- Packaging:Plastic vial
Rabbit polyclonal antibody to ERD22 for WB, IF and ELISA with samples derived from Human, Mouse and Rat.
- Validated Applications: WB, IF, ELISA
- Recommended Dilutions: WB: 1:500-1:1000, IF: 1:100-1:500, ELISA: 1:20000
Type: Primary
Antigen: KDELR2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat