Anti-Shugoshin Rabbit Polyclonal Antibody
Catalog # 77052-444
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:hSgo 1
- Antigen Symbol:Shugoshin
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q5FBB7
- Antigen Synonyms:SGOL 1|Serologically defined breast cancer antigen NY BR 85|NY BR 85|SGO L1|hSgo1|SGO|SGOL1_HUMAN|SGOL1|Serologically defined breast cancer antigen NY-BR-85
- Storage Buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular Weight:64 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human SGO1 around the phosphorylation site of Ser14 (amino acids 1-50)
- Tested Applications:IHC
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing phospho peptide
- Cat. No.:77052-444
- Packaging:Plastic vial
Specifications
About this item
Rabbit polyclonal antibody to SGO1 (phospho Ser14) for IHC and ELISA with samples derived from Human.
- Validated Applications: IHC, ELISA
- Recommended Dilutions: IHC: 1:50-1:100, ELISA: 1:10000
Type: Primary
Antigen: Shugoshin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human