Anti-HIC5 Rabbit Polyclonal Antibody
77052-004
- Antibody Type:Primary
- Antigen Name:Androgen receptor coactivator 55 kDa protein
- Antigen Symbol:HIC5
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# O43294
- Antigen Synonyms:Transforming growth factor beta 1 induced transcript 1|Hic-5|Androgen receptor coactivator ARA55|HIC5|TGFI1_HUMAN|Transforming growth factor beta-1-induced transcript 1 protein|Hydrogen peroxide-inducible clone 5 protein|Androgen receptor-associated protein of 55 kDa|Tgfb1i1
- Storage Buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular Weight:49 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human Hic-5 (amino acids 31-80)
- Tested Applications:IHC
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Cat. No.:77052-004
- Packaging:Plastic vial
Rabbit polyclonal antibody to Hic-5 for IHC, IF and ELISA with samples derived from Human, Mouse and Rat.
- Validated Applications: IHC, IF, ELISA
- Recommended Dilutions: IHC: 1:50-1:100, IF: 1:100-1:500, ELISA: 1:1000
Type: Primary
Antigen: HIC5
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat