Order Entry
United States
ContactUsLinkComponent
 
Anti-MMP17 Rabbit Polyclonal Antibody
  77051-180
undefined
Anti-MMP17 Rabbit Polyclonal Antibody
  77051-180
 :  A95658-100

 

  • Antibody Type:
    Primary
  • Antigen Name:
    Matrix metalloproteinase 17
  • Antigen Symbol:
    MMP17
  • Clonality:
    Polyclonal
  • Conjugation:
    Unconjugated
  • ELISA:
    Yes
  • Host:
    Rabbit
  • Isotype:
    IgG
  • Reactivity:
    Human
  • Western Blot:
    Yes
  • Size:
    100 µg
  • Form:
    Liquid
  • Gene ID:
    UniprotID# Q9ULZ9
  • Antigen Synonyms:
    Membrane type matrix metalloproteinase 4|Membrane-type-4 matrix metalloproteinase|MMP 17|MMP-17|MMP17|Membrane type 4 matrix metalloproteinase|Membrane-type matrix metalloproteinase 4|Matrix metalloproteinase 17 membrane inserted|Matrix metalloproteinase-17
  • Storage Buffer:
    Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
  • Molecular Weight:
    53 kDa
  • Storage Temperature:
    Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
  • Shipping Temperature:
    Shipped on blue ice at 4 °C
  • Immunogen:
    Synthetic peptide derived from human MMP17 (amino acids 110-159)
  • Purification:
    Purified from rabbit serum by antigen affinity chromatography using the immunizing phospho peptide
  • Cat. No.:
    77051-180
  • Packaging:
    Plastic vial

 

 

Rabbit polyclonal antibody to MMP17 (cleaved Gln129) for WB and ELISA with samples derived from Human.

  • Validated Applications: WB, ELISA
  • Recommended Dilutions: WB: 1:500-1:1000, ELISA: 1:1000

Type: Primary
Antigen: MMP17
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human