Anti-OR4F4 + OR4F5 + OR4F17 Rabbit Polyclonal Antibody
77051-156
- Antibody Type:Primary
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q8NGA8, Q96R69, Q8NH21
- Storage Buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular Weight:34 kDa
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human OR4F4/4F5/4F17 (amino acids 51-100)
- Purification:Immunogen affinity purified
- Cat. No.:77051-156
- Packaging:Plastic vial
Rabbit polyclonal antibody to OR4F4 + OR4F5 + OR4F17 for WB and ELISA with samples derived from Human.
- Validated Applications: WB, ELISA
Type: Primary
Antigen:
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human