Anti-ATP6V0D1 Rabbit Polyclonal Antibody
77050-704
- Antibody Type:Primary
- Antigen Name:32 kDa accessory protein
- Antigen Symbol:ATP6V0D1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P61421
- Antigen Synonyms:ATPase H+ transporting lysosomal (vacuolar proton pump) member D|ATP6V0D1|ATPase H+ transporting lysosomal V0 subunit d1|ATPase H+ transporting lysosomal 38kDa V0 subunit d1|ATP6DV|p39|ATP6D|H(+) transporting two sector ATPase subunit D|ATPase H+ transporting lysosomal 38kD V0 subunit d
- Storage Buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular Weight:40 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human V-ATPase D1 (amino acids 221-270)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Cat. No.:77050-704
- Packaging:Plastic vial
Rabbit polyclonal antibody to V-ATPase D1 for WB and ELISA with samples derived from Human, Mouse and Rat.
- Validated Applications: WB, ELISA
- Recommended Dilutions: WB: 1:500-1:1000, ELISA: 1:10000
Type: Primary
Antigen: ATP6V0D1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat