Anti-SGK1 Rabbit Polyclonal Antibody
77050-448
- Antibody Type:Primary
- Antigen Name:OTTHUMP00000017247
- Antigen Symbol:SGK1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# O00141
- Antigen Synonyms:Serine/threonine protein kinase Sgk1|Serum/glucocorticoid regulated kinase 1|Sgk1 variant i3|SGK 1|Serum and glucocorticoid regulated kinase|Serine/threonine protein kinase SGK|Serum/glucocorticoid-regulated kinase 1|Serum/glucocorticoid regulated kinase|Serine/threonine-protein kinase Sgk1
- Storage Buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular Weight:48 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human SGK (amino acids 41-90)
- Tested Applications:IHC
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Cat. No.:77050-448
- Packaging:Plastic vial
Rabbit polyclonal antibody to SGK for WB, IHC, IF and ELISA with samples derived from Human, Mouse and Rat.
- Validated Applications: WB, IHC, IF, ELISA
- Recommended Dilutions: WB: 1:500-1:1000, IHC: 1:50-1:100, IF: 1:100-1:500, ELISA: 1:5000
Type: Primary
Antigen: SGK1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat