Anti-5HT2C Receptor Rabbit Polyclonal Antibody
77050-416
- Antibody Type:Primary
- Antigen Name:5 Hydroxytryptamine 2C receptor
- Antigen Symbol:5HT2C Receptor
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P28335
- Antigen Synonyms:5-HT-1C|5-ht-1c receptor|5-hydroxytryptamine receptor 2C|5-HT1C|5-HTR2C|5-HT-2C|5-hydroxytryptamine receptor 1C|5-hydroxytryptamine (serotonin) receptor 2C, G protein-coupled|5-HT2C
- Storage Buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular Weight:51 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human 5-HT-2C (amino acids 161-210)
- Tested Applications:IHC
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Cat. No.:77050-416
- Packaging:Plastic vial
Rabbit polyclonal antibody to 5-HT-2C for IHC, IF and ELISA with samples derived from Human, Mouse and Rat.
- Validated Applications: IHC, IF, ELISA
- Recommended Dilutions: IHC: 1:50-1:100, IF: 1:100-1:500, ELISA: 1:5000
Type: Primary
Antigen: 5HT2C Receptor
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat