Anti-p63 Protein + HMW Cytokeratin Rabbit Polyclonal Antibody
77049-668
- Antibody Type:Primary
- Antigen Name:CK13
- Antigen Symbol:p63 Protein + HMW Cytokeratin
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q9H3D4
- Antigen Synonyms:Cytokeratin 13|KRT13|Cytokeratin 15|KRT14|Cytokeratin 14|Cytokeratin 16|CK16|CK14|CK15
- Storage Buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular Weight:76 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human p63 around the phosphorylation site of Ser455 (amino acids 421-470)
- Tested Applications:IHC
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing phospho peptide
- Cat. No.:77049-668
- Packaging:Plastic vial
Rabbit polyclonal antibody to p63 (phospho Ser455) for WB, IHC and ELISA with samples derived from Human.
- Validated Applications: WB, IHC, ELISA
- Recommended Dilutions: WB: 1:500-1:1000, IHC: 1:50-1:100, ELISA: 1:20000
Type: Primary
Antigen: p63 Protein + HMW Cytokeratin
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human