Anti-Prostaglandin I2 Receptor Rabbit Polyclonal Antibody
Catalog # 77049-648
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:Prostaglandin I2 (prostacyclin) receptor (IP)
- Antigen Symbol:Prostaglandin I2 Receptor
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P43119
- Antigen Synonyms:PI2R_HUMAN|PGI receptor|Prostaglandin I2 Receptor|Ptgir|Prostacyclin receptor|IP|PGI2 receptor|Prostanoid IP receptor|PRIPR
- Storage Buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular Weight:40 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human Prostacyclin Receptor (amino acids 198-247)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Cat. No.:77049-648
- Packaging:Plastic vial
Specifications
About this item
Rabbit polyclonal antibody to Prostacyclin Receptor for WB, IF and ELISA with samples derived from Human.
- Validated Applications: WB, IF, ELISA
- Recommended Dilutions: WB: 1:500-1:1000, IF: 1:100-1:500, ELISA: 1:10000
Type: Primary
Antigen: Prostaglandin I2 Receptor
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human