Anti-Chk1 Rabbit Polyclonal Antibody
Catalog # 77049-066
Supplier: ANTIBODIES.COM LLC
Specifications
- Antibody Type:Primary
- Antigen Name:C85740
- Antigen Symbol:Chk1
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# O14757
- Antigen Synonyms:CHK1 checkpoint homolog (S. pombe)|CHEK 1|Chk 1|Cell cycle checkpoint kinase|Chek1|Chk1|Checkpoint , S. pombe, homolog of, 1|Checkpoint kinase 1|Checkpoint kinase 1 homolog (S. pombe)
- Storage Buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular Weight:54 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human Chk1 around the phosphorylation site of Ser296 (amino acids 266-315)
- Tested Applications:IHC
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing phospho peptide
- Cat. No.:77049-066
- Packaging:Plastic vial
Specifications
About this item
Rabbit polyclonal antibody to Chk1 (phospho Ser296) for WB, IHC and ELISA with samples derived from Human.
- Validated Applications: WB, IHC, ELISA
- Recommended Dilutions: WB: 1:500-1:1000, IHC: 1:50-1:100, ELISA: 1:5000
Type: Primary
Antigen: Chk1
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human