Anti-APRIL Rabbit Polyclonal Antibody
77048-908
- Antibody Type:Primary
- Antigen Name:A proliferation inducing ligand
- Antigen Symbol:APRIL
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# O75888
- Antigen Synonyms:TNF-related death ligand 1|A proliferation-inducing ligand|APRIL|TNF- and APOL-related leukocyte expressed ligand 2|TALL2|TALL-2|TNF related death ligand|CD256|TNF13_HUMAN
- Storage Buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular Weight:27 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human TALL-2 (amino acids 151-200)
- Tested Applications:IHC
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Cat. No.:77048-908
- Packaging:Plastic vial
Rabbit polyclonal antibody to TALL-2 for WB, IHC, IF and ELISA with samples derived from Human, Mouse and Rat.
- Validated Applications: WB, IHC, IF, ELISA
- Recommended Dilutions: WB: 1:500-1:1000, IHC: 1:50-1:100, IF: 1:100-1:500, ELISA: 1:10000
Type: Primary
Antigen: APRIL
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat