Anti-TRA2A Rabbit Polyclonal Antibody
77048-776
- Antibody Type:Primary
- Antigen Name:AWMS1
- Antigen Symbol:TRA2A
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q13595
- Antigen Synonyms:Tra2alpha|TRA 2 alpha|TRA 2A|TRA2 alpha|htra-2 alpha|TRA2A|TRA2-alpha|HSU53209|Putative MAPK activating protein PM24
- Storage Buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular Weight:32 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human TRA-2 alpha (amino acids 221-270)
- Tested Applications:IHC
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Cat. No.:77048-776
- Packaging:Plastic vial
Rabbit polyclonal antibody to TRA-2 alpha for WB, IHC, IF and ELISA with samples derived from Human and Mouse.
- Validated Applications: WB, IHC, IF, ELISA
- Recommended Dilutions: WB: 1:500-1:1000, IHC: 1:50-1:100, IF: 1:100-1:500, ELISA: 1:20000
Type: Primary
Antigen: TRA2A
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse