Anti-pan Synuclein Rabbit Polyclonal Antibody
77048-492
- Antibody Type:Primary
- Antigen Name:BCSG1
- Antigen Symbol:pan Synuclein
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Rat,Mouse
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# P37840
- Antigen Synonyms:pan Synuclein|PARK1|Persyn|SNCA|Breast cancer specific gene 1 protein|PRSN|Gamma synuclein|beta synuclein|NACP
- Storage Buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular Weight:14 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic phospho-peptide derived from human Synuclein-alpha around the phosphorylation site of Tyr136
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing peptide
- Cat. No.:77048-492
- Packaging:Plastic vial
Rabbit polyclonal antibody to Synuclein-alpha (phospho Tyr136) for WB and ELISA with samples derived from Human, Mouse and Rat.
- Validated Applications: WB, ELISA
- Recommended Dilutions: WB: 1:500-1:1000, ELISA: 1:10000
Type: Primary
Antigen: pan Synuclein
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse, Rat