Anti-ITK Rabbit Polyclonal Antibody
77048-308
- Antibody Type:Primary
- Antigen Name:EMT
- Antigen Symbol:ITK
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- ImmunoChemistry:Yes
- Isotype:IgG
- Reactivity:Human,Mouse
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# Q08881
- Antigen Synonyms:ITK_HUMAN|Homolog of mouse T cell itk/tsk|IL 2 inducible T cell kinase|Kinase EMT|IL2 inducible T cell kinase|LPFS1|Interleukin 2 inducible T cell kinase|LYK|ITK
- Storage Buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular Weight:71 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human ITK around the phosphorylation site of Tyr512 (amino acids 478-527)
- Tested Applications:IHC
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing phospho peptide
- Cat. No.:77048-308
- Packaging:Plastic vial
Rabbit polyclonal antibody to ITK (phospho Tyr512) for IHC and ELISA with samples derived from Human and Mouse.
- Validated Applications: IHC, ELISA
- Recommended Dilutions: IHC: 1:50-1:100, ELISA: 1:10000
Type: Primary
Antigen: ITK
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse