Anti-Ephrin A2 Rabbit Polyclonal Antibody
77047-824
- Antibody Type:Primary
- Antigen Name:EFNA 2
- Antigen Symbol:Ephrin A2
- Clonality:Polyclonal
- Conjugation:Unconjugated
- ELISA:Yes
- Host:Rabbit
- Isotype:IgG
- Reactivity:Human,Mouse
- Western Blot:Yes
- Size:100 µg
- Form:Liquid
- Gene ID:UniprotID# O43921
- Antigen Synonyms:ELF1|EFNA2_HUMAN|EPLG6|Ephrin A2|Eph related receptor tyrosine kinase ligand 6|Ephrin-A2|EPH-related receptor tyrosine kinase ligand 6|EphrinA2|efna2
- Storage Buffer:Supplied in Phosphate Buffered Saline (without Mg²⁺ and Ca²⁺), pH 7.4, with 150 mM NaCl, 0.02% Sodium Azide, and 50% Glycerol
- Molecular Weight:21 kDa
- Storage Temperature:Shipped at 4 °C. Upon delivery aliquot and store at −20 °C. Avoid freeze/thaw cycles.
- Shipping Temperature:Shipped on blue ice at 4 °C
- Immunogen:Synthetic peptide derived from human EFNA2 (amino acids 139-188)
- Purification:Purified from rabbit serum by antigen affinity chromatography using the immunizing phospho peptide
- Cat. No.:77047-824
- Packaging:Plastic vial
Rabbit polyclonal antibody to EFNA2 (cleaved Asn188) for WB and ELISA with samples derived from Human and Mouse.
- Validated Applications: WB, ELISA
- Recommended Dilutions: WB: 1:500-1:1000, ELISA: 1:10000
Type: Primary
Antigen: Ephrin A2
Clonality: Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human, Mouse